Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033274.4 | NP_150377.1 | 19, 20 | 2363 | ||
XM_005266003.2 | XP_005266060.1 | 19, 20 | 2315 | |||
XM_011534682.2 | XP_011532984.1 | 12, 13 | 3119 | |||
XM_017010009.1 | XP_016865498.1 | 12, 13 | 1580 | |||
GenBank mRNA | AF134707.1 | - | 16, 17 | 1944 | ||
AF311317.1 | - | 19, 20 | 2272 | |||
AF326918.1 | - | 19, 20 | 2284 | |||
AK091464.1 | - | 19, 20 | 2325 | |||
AK308182.1 | - | 19, 20 | 2315 | |||
BC024214.2 | - | 8, 9 | 948 | |||
BC033132.1 | - | 8, 9 | 948 | |||
Y13786.2 | - | 19, 20 | 2313 |