Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006056.4 | NP_006047.3 | 3 | 1509 | ||
XM_011510487.2 | XP_011508789.1 | 3 | 1497 | |||
GenBank mRNA | AK300222.1 | - | 3 | 1532 | ||
BC036543.1 | - | 3 | 1537 | 5265972 | ||
BC051914.1 | - | 3 | 1463 | 6194542 |
Set Membership: |
G-protein coupled receptor |