Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018490.2 | NP_060960.2 | 18 | 2941 | ||
GenBank mRNA | AB209743.1 | - | 18 | 2860 | ||
AF257182.1 | - | 18 | 2900 | |||
BC033039.2 | - | 17 | 2857 | 5260923 |