Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK2N1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018584.5 | NP_061054.2 | 1, 2 | 1026 | ||
GenBank mRNA | AY204901.1 | - | 1, 2 | 570 | ||
BC020630.1 | - | 12 |