Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183374.2 | NP_899230.2 | 6 | 1225 | ||
GenBank mRNA | AK127234.1 | - | 2 | 1741 | ||
AK131099.1 | - | 6 | 3026 | |||
AY356349.1 | - | 6 | 1225 |