Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 3 | 780 | ||
NM_012470.3 | NP_036602.1 | 3 | 780 | |||
NR_034053.2 | 4 | 941 | ||||
XM_011515989.2 | XP_011514291.1 | 3 | 382 | |||
GenBank mRNA | AF145029.1 | - | 3 | 490 | ||
AJ133769.1 | - | 3 | 377 | |||
AK225999.1 | - | 3 | 751 | |||
AK312580.1 | - | 3 | 507 | |||
BC068530.1 | - | 3 | 508 |