Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013242.2 | NP_037374.1 | 2, 3 | 436 | ||
NR_136632.1 | 2, 3 | 469 | ||||
GenBank mRNA | AF087845.1 | - | 2, 3 | 416 | ||
AF093680.1 | - | 2, 3 | 415 | |||
AK027099.1 | - | 2, 3 | 357 | |||
AK297819.1 | - | 2, 3 | 391 | |||
AV751497.1 | - | 2, 3 | 436 | |||
BC005152.2 | - | 2, 3 | 405 | 3528301 | ||
CN364433.1 | - | 2, 3 | 396 |