Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTNNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127384.2 | NP_001120856.1 | 3 | 347 | ||
NM_001291133.1 | NP_001278062.1 | 4 | 424 | |||
NM_013266.3 | NP_037398.2 | 3 | 394 | |||
XM_017016151.1 | XP_016871640.1 | 3 | 320 | |||
XM_017016152.1 | XP_016871641.1 | 4 | 497 | |||
XM_017016153.1 | XP_016871642.1 | 4 | 397 | |||
XM_017016158.1 | XP_016871647.1 | 4 | 495 | |||
GenBank mRNA | AF091606.1 | - | 3 | 372 | ||
AK289756.1 | - | 3 | 283 | |||
AK308726.1 | - | 3 | 261 | |||
AK313882.1 | - | 3 | 397 | |||
AY198183.1 | - | 4 | 363 | |||
BC022004.2 | - | 4 | 419 | |||
BC030646.2 | - | 4 | 419 | |||
BC065819.1 | - | 3 | 290 | 6189390 | ||
BP362015.1 | - | 3 | 320 |