Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other POLL SIRNA Assays ›
Gene Symbol:
POLL
Gene Name:
polymerase (DNA) lambda
Gene Aliases:
BETAN, POLKAPPA
Chromosome Location:
Chr.10: 101578882 - 101588270 on Build GRCh38
UniGene ID:
Hs.523230
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
20282
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: POLL
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001174084.1
NP_001167555.1
3
738
NM_001174085.1
NP_001167556.1
3
675
NM_013274.3
NP_037406.1
3
675
NR_033406.1
3
274
XM_011539650.1
XP_011537952.1
3
343
XM_011539651.1
XP_011537953.1
3
343
XM_011539652.1
XP_011537954.1
3
631
XM_011539653.1
XP_011537955.1
3
624
XM_011539654.1
XP_011537956.1
3
343
XM_011539655.1
XP_011537957.1
3
343
XM_011539656.1
XP_011537958.1
3
343
XM_011539657.1
XP_011537959.1
3
570
XM_011539667.1
XP_011537969.1
3
305
XM_017016084.1
XP_016871573.1
3
343
XM_017016085.1
XP_016871574.1
3
343
XM_017016086.1
XP_016871575.1
2
1051
XM_017016087.1
XP_016871576.1
3
343
XM_017016088.1
XP_016871577.1
3
343
XM_017016090.1
XP_016871579.1
3
343
XM_017016092.1
XP_016871581.1
3
624
GenBank mRNA
AF161019.1
-
2
304
AF283478.1
-
2
243
AJ131890.1
-
3
614
AK021600.1
-
3
346
AK022476.1
-
3
252
AK292225.1
-
3
597
AK293421.1
-
3
335
AK293657.1
-
2
180
AY302442.1
-
2
243
BC068529.1
-
3
627
5267859
DA107389.1
-
3
274
DC379162.1
-
3
274
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-directed DNA polymerase
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
nucleotide-excision repair
double-strand break repair via nonhomologous end joining
somatic hypermutation of immunoglobulin genes
DNA biosynthetic process
DNA binding
DNA-directed DNA polymerase activity
lyase activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-k7c2b:80/10.239.160.46:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0