Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191028.2 | NP_001177957.2 | 20 | 2851 | ||
NM_012470.3 | NP_036602.1 | 21 | 3043 | |||
NR_034053.2 | 22 | 3204 | ||||
XM_011515989.2 | XP_011514291.1 | 21 | 2645 | |||
GenBank mRNA | AF145029.1 | - | 21 | 2855 | ||
AJ133769.1 | - | 21 | 2640 | |||
AK001263.1 | - | 14 | 1597 | |||
AK022910.1 | - | 12 | 1377 | |||
AK225999.1 | - | 20 | 2822 | |||
AK312580.1 | - | 21 | 2770 | |||
BC009221.2 | - | 9 | 887 | |||
BC068530.1 | - | 21 | 2771 |