Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CLCF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166212.1 | NP_001159684.1 | 3 | 1569 | ||
NM_013246.2 | NP_037378.1 | 3 | 1650 | |||
GenBank mRNA | AF172854.1 | - | 3 | 1499 | ||
BC012939.1 | - | 3 | 1526 | 4453813 |