Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114094.1 | NP_001107566.1 | 16 | 1397 | ||
NM_001258440.1 | NP_001245369.1 | 16 | 1250 | |||
NM_001258441.1 | NP_001245370.1 | 15 | 1181 | |||
NM_001258442.1 | NP_001245371.1 | 13 | 935 | |||
NM_013314.3 | NP_037446.1 | 17 | 1466 | |||
NR_047680.1 | 16 | 1230 | ||||
NR_047681.1 | 15 | 1178 | ||||
NR_047682.1 | 15 | 1173 | ||||
NR_047683.1 | 14 | 1111 | ||||
XM_011539728.2 | XP_011538030.1 | 17 | 1343 | |||
XM_017016159.1 | XP_016871648.1 | 16 | 1275 | |||
GenBank mRNA | AF068180.1 | - | 17 | 1441 | ||
AF068181.1 | - | 16 | 1372 | |||
AK027779.1 | - | 17 | 1401 | |||
AK225546.1 | - | 16 | 1380 | |||
AM180327.1 | - | 13 | 817 | |||
AM180328.1 | - | 7 | 472 | |||
AM180329.1 | - | 5, 6 | 317 | |||
AM180331.1 | - | 7 | 465 | |||
AM180332.1 | - | 15 | 1063 | |||
AM180333.1 | - | 16 | 1219 | |||
AM180334.1 | - | 16 | 1219 | |||
AM180335.1 | - | 13 | 857 | |||
AM180337.1 | - | 16 | 1132 | |||
AM180338.1 | - | 15 | 1060 | |||
AM180339.1 | - | 15 | 1055 | |||
AM180340.1 | - | 14 | 993 | |||
AM180341.1 | - | 5 | 330 | |||
AM180342.1 | - | 15 | 1082 | |||
AM180343.1 | - | 16 | 1238 | |||
AM180344.1 | - | 14 | 1128 | |||
AM180345.1 | - | 13 | 943 | |||
AM180346.1 | - | 14 | 1155 | |||
BC018906.2 | - | 17 | 1380 | 4053894 | ||
BC110298.1 | - | 16 | 1222 | |||
BF975580.1 | - | 5 | 397 |