Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNPO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136195.1 | NP_001129667.1 | 5 | 591 | ||
NM_001136196.1 | NP_001129668.1 | 4 | 418 | |||
NM_013433.4 | NP_038461.2 | 4 | 404 | |||
GenBank mRNA | AB210011.1 | - | 3 | 971 | ||
AF007748.1 | - | 5 | 524 | |||
AF019039.1 | - | 4 | 357 | |||
AK299485.1 | - | 4 | 331 | |||
BC018314.1 | - | 4 | 295 | |||
BC072420.1 | - | 4 | 377 | 6184325 | ||
DA100622.1 | - | 4 | 419 | |||
DA808781.1 | - | 4 | 468 |