Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287242.1 | NP_001274171.1 | 4 | 533 | ||
NM_006125.2 | NP_006116.2 | 4 | 1756 | |||
NM_013423.2 | NP_038267.1 | 4 | 585 | |||
NM_013427.2 | NP_038286.2 | 4 | 1756 | |||
NR_109776.1 | 5 | 1852 | ||||
GenBank mRNA | AB208792.1 | - | 4 | 1138 | ||
AF012272.2 | - | 4 | 1757 | |||
AF022212.2 | - | 5 | 1853 | |||
AF117067.1 | - | 4 | 1757 | |||
AF177663.1 | - | 4 | 585 | |||
AF177665.1 | - | 4 | 391 | |||
AK297713.1 | - | 4 | 430 | |||
AK303444.1 | - | 4 | 446 | |||
BC150635.1 | - | 4 | 915 | 9057005 |