Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013393.1 | NP_037525.1 | 3 | 779 | ||
GenBank mRNA | AF093415.1 | - | 3 | 779 | ||
AK024635.1 | - | 2 | 528 | |||
AK302060.1 | - | 4 | 890 | |||
BC017106.2 | - | 2 | 742 | |||
BC114514.1 | - | 3 | 764 | 40031660 | ||
BC114564.1 | - | 3 | 733 | |||
CR749618.1 | - | 3 | 788 | |||
EU794658.1 | - | 3 | 779 |