Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085454.1 | NP_001078923.1 | 15 | 1764 | ||
NM_014030.3 | NP_054749.2 | 14 | 1737 | |||
XM_011524684.1 | XP_011522986.1 | 15 | 1605 | |||
XM_011524685.1 | XP_011522987.1 | 14 | 1578 | |||
GenBank mRNA | AB209537.1 | - | 15 | 1563 | ||
AF124490.1 | - | 14 | 1537 | |||
AK294785.1 | - | 16 | 1642 | |||
AK297443.1 | - | 12 | 1206 | |||
AK299615.1 | - | 6 | 634 | |||
AK299932.1 | - | 15 | 1608 | |||
BC067358.1 | - | 14 | 1737 | |||
BX537553.1 | - | 7 | 1313 | |||
BX537554.1 | - | 7 | 1313 | |||
BX648097.1 | - | 7 | 1315 |