Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014043.3 | NP_054762.2 | 2 | 335 | ||
XM_011533576.2 | XP_011531878.1 | 2 | 166 | |||
GenBank mRNA | AF151842.1 | - | 2 | 294 | ||
AK002165.1 | - | 2 | 308 | |||
AK002180.1 | - | 2 | 313 | |||
AK222654.1 | - | 2 | 295 | |||
AK316585.1 | - | 2 | 299 | |||
AL080122.1 | - | 2 | 308 | |||
BC001553.1 | - | 2 | 151 | 3460712 | ||
BI821848.1 | - | 2 | 312 | |||
BM828410.1 | - | 2 | 335 | |||
CR533456.1 | - | 2 | 75 | |||
DA426159.1 | - | 2 | 306 |