Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIGD1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099668.1 | NP_001093138.1 | 3 | 506 | ||
NM_001099669.1 | NP_001093139.1 | 3 | 359 | |||
NM_014056.3 | NP_054775.2 | 3 | 335 | |||
GenBank mRNA | AF077034.1 | - | 3 | 287 | ||
AF145385.1 | - | 3 | 274 | |||
AK225461.1 | - | 3 | 259 | |||
AK225571.1 | - | 3 | 290 | |||
AK311815.1 | - | 3 | 290 | |||
AL110233.1 | - | 3 | 305 | |||
BC000601.1 | - | 3 | 273 | 3346355 | ||
BC009583.1 | - | 3 | 266 | 4093421 | ||
BC009594.1 | - | 3 | 272 | 4109190 | ||
BC070277.1 | - | 3 | 288 | 4686431 | ||
BG708126.1 | - | 3 | 340 | |||
BP208784.1 | - | 3 | 505 | |||
BP370118.1 | - | 3 | 281 | |||
CR457385.1 | - | 2 | 213 |