Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014043.3 | NP_054762.2 | 2 | 345 | ||
XM_011533576.2 | XP_011531878.1 | 2 | 176 | |||
GenBank mRNA | AF151842.1 | - | 2 | 304 | ||
AK002165.1 | - | 2 | 318 | |||
AK002180.1 | - | 2 | 323 | |||
AK222654.1 | - | 2 | 305 | |||
AK316585.1 | - | 2 | 309 | |||
AL080122.1 | - | 2 | 318 | |||
BC001553.1 | - | 2 | 161 | 3460712 | ||
BI821848.1 | - | 2 | 322 | |||
BM828410.1 | - | 2 | 345 | |||
CR533456.1 | - | 2 | 85 | |||
DA426159.1 | - | 2 | 316 |