Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271707.1 | NP_001258636.1 | 2 | 274 | ||
NM_014160.4 | NP_054879.3 | 3 | 403 | |||
GenBank mRNA | AF302084.1 | - | 3 | 317 | ||
AK091318.1 | - | 3 | 348 | |||
AK298463.1 | - | 2 | 201 | |||
BC001799.2 | - | 1 | 112 | |||
BC015715.1 | - | 3 | 345 | 3916199 | ||
DR157266.1 | - | 3 | 403 |