Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308142.1 | NP_001295071.1 | 17 | 8019 | ||
NM_014048.4 | NP_054767.3 | 17 | 7870 | |||
XM_005255452.3 | XP_005255509.1 | 18 | 8174 | |||
XM_005255453.4 | XP_005255510.1 | 17 | 8658 | |||
XM_005255455.3 | XP_005255512.1 | 15 | 8004 | |||
XM_006720908.3 | XP_006720971.1 | 18 | 8820 | |||
XM_006720909.3 | XP_006720972.1 | 18 | 8149 | |||
XM_006720912.3 | XP_006720975.1 | 15 | 9959 | |||
XM_006720913.2 | XP_006720976.1 | 17 | 8036 | |||
XM_006720914.2 | XP_006720977.1 | 18 | 8024 | |||
XM_011522568.2 | XP_011520870.1 | 17 | 7989 | |||
XM_011522569.2 | XP_011520871.1 | 15 | 7786 | |||
XM_017023501.1 | XP_016878990.1 | 17 | 7985 | |||
XM_017023502.1 | XP_016878991.1 | 17 | 8042 | |||
XM_017023503.1 | XP_016878992.1 | 16 | 11187 | |||
XM_017023504.1 | XP_016878993.1 | 15 | 7854 | |||
XM_017023505.1 | XP_016878994.1 | 15 | 9809 | |||
GenBank mRNA | AB033069.2 | - | 9 | 7026 | ||
AF078853.1 | - | 1 | 772 | |||
AK056385.1 | - | 1 | 2352 | |||
AK057216.1 | - | 1 | 1386 | |||
BC006977.1 | - | 1 | 571 | |||
BC016293.1 | - | 1 | 794 | |||
BC020644.1 | - | 1 | 499 | |||
BC030510.1 | - | 1 | 664 | |||
BC050541.1 | - | 1 | 494 | |||
BC107868.1 | - | 1 | 799 | |||
BX641041.1 | - | 5 | 7107 | |||
CR749797.1 | - | 6 | 6565 |