Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCOA6 SIRNA Assays ›
Gene Symbol:
NCOA6
Gene Name:
nuclear receptor coactivator 6
Gene Aliases:
AIB3, ASC2, NRC, PRIP, RAP250, TRBP
Chromosome Location:
Chr.20: 34714774 - 34825649 on Build GRCh38
UniGene ID:
Hs.368971
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
20888
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NCOA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242539.2
NP_001229468.1
7
2314
NM_001318240.1
NP_001305169.1
10
4836
NM_014071.4
NP_054790.2
9
2606
XM_006723755.3
XP_006723818.1
9
2534
XM_011528721.2
XP_011527023.1
9
2551
XM_011528726.2
XP_011527028.1
9
2518
XM_011528727.2
XP_011527029.1
9
2515
XM_017027739.1
XP_016883228.1
10
2608
XM_017027740.1
XP_016883229.1
11
2727
XM_017027741.1
XP_016883230.1
10
2608
XM_017027742.1
XP_016883231.1
10
2593
XM_017027743.1
XP_016883232.1
10
2608
XM_017027744.1
XP_016883233.1
9
2497
XM_017027745.1
XP_016883234.1
9
2464
XM_017027746.1
XP_016883235.1
10
2608
XM_017027747.1
XP_016883236.1
7
1985
XM_017027748.1
XP_016883237.1
10
2608
XM_017027749.1
XP_016883238.1
10
2608
XM_017027750.1
XP_016883239.1
10
2608
XM_017027751.1
XP_016883240.1
9
2534
GenBank mRNA
AF128458.1
-
8
2467
AF171667.1
-
8
2409
AF177388.1
-
8
2425
AF208227.1
-
10
4833
AF245115.1
-
7
2324
BC136272.1
-
8
2415
9020259
BU187688.1
-
1
313
D80003.2
-
8
2402
JF707637.1
-
7
2265
JF707638.1
-
7
2265
JF707639.1
-
7
2265
Back To Top
More Information
Panther Classification:
Molecular Function -
chromatin/chromatin-binding, or -regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
DNA replication
DNA repair
DNA recombination
DNA-templated transcription, initiation
transcription initiation from RNA polymerase II promoter
cellular response to DNA damage stimulus
brain development
heart development
response to hormone
myeloid cell differentiation
intracellular estrogen receptor signaling pathway
positive regulation of insulin secretion involved in cellular response to glucose stimulus
glucocorticoid receptor signaling pathway
cellular lipid metabolic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
chromatin binding
transcription coactivator activity
protein binding
ligand-dependent nuclear receptor binding
enzyme binding
protein domain specific binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
glucocorticoid receptor binding
protein homodimerization activity
peroxisome proliferator activated receptor binding
retinoid X receptor binding
thyroid hormone receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-46vz9:80/10.239.163.144:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0