Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMARCAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127207.1 | NP_001120679.1 | 12, 13 | 2213 | ||
NM_014140.3 | NP_054859.2 | 12, 13 | 2399 | |||
XM_005246631.2 | XP_005246688.1 | 12, 13 | 2337 | |||
XM_005246632.1 | XP_005246689.1 | 12, 13 | 2389 | |||
XM_017004228.1 | XP_016859717.1 | 12, 13 | 2202 | |||
GenBank mRNA | AF082179.1 | - | 11, 12 | 2169 | ||
AF432223.1 | - | 12, 13 | 2209 | |||
AK000117.1 | - | 8, 9 | 1069 | |||
AK023780.1 | - | 12, 13 | 2339 | |||
AL122076.1 | - | 10, 11 | 1633 | |||
BC016482.1 | - | 11, 12 | 2160 | 4341673 | ||
BC043341.1 | - | 12, 13 | 2186 | 6058575 |