Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014146.3 | NP_054865.2 | 13 | 1398 | ||
NM_032463.2 | NP_115852.1 | 14 | 1517 | |||
NM_032464.2 | NP_115853.2 | 14 | 1587 | |||
XM_011516558.2 | XP_011514860.1 | 14 | 1427 | |||
GenBank mRNA | AF161531.1 | - | 13 | 1148 | ||
AF252611.1 | - | 14 | 1338 | |||
AF252612.1 | - | 12 | 1093 | |||
AF252613.1 | - | 13 | 1134 | |||
AF252614.1 | - | 14 | 1268 | |||
AF257135.1 | - | 12 | 1069 | |||
AK002099.1 | - | 14 | 1377 | |||
AK092904.1 | - | 14 | 1728 | |||
AK225150.1 | - | 12 | 834 | |||
BC001609.1 | - | 14 | 1399 | 3543310 | ||
BC009204.2 | - | 12 | 1071 | 4054137 |