Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGA9-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110532.1 | 6 | 496 |
Gene Symbol: NDUFAF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014165.3 | NP_054884.1 | 3 | 432 | ||
GenBank mRNA | AF060508.1 | - | 3 | 378 | ||
AF161474.1 | - | 3 | 421 | |||
AK026245.1 | - | 2 | 277 | |||
AK311850.1 | - | 3 | 405 | |||
BC018802.1 | - | 3 | 418 | |||
BC039464.1 | - | 3 | 411 | 4388671 | ||
DB253189.1 | - | 3 | 432 |