Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014248.3 | NP_055063.1 | 2 | 197 | ||
GenBank mRNA | AF085906.1 | - | 2 | 126 | ||
AF140598.1 | - | 2 | 151 | |||
AF142059.1 | - | 2 | 145 | |||
AK315722.1 | - | 2 | 156 | |||
AY099360.1 | - | 2 | 108 | |||
BC001466.2 | - | 2 | 150 | 3138751 | ||
BC017370.1 | - | 2 | 167 | |||
CR456560.1 | - | 2 | 163 | |||
DA353856.1 | - | 2 | 197 |