Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278000.1 | NP_001264929.1 | 5 | 1199 | ||
NM_005296.2 | NP_005287.1 | 2 | 984 | |||
XM_005262126.3 | XP_005262183.1 | 5 | 1402 | |||
XM_006724639.3 | XP_006724702.1 | 4 | 1114 | |||
XM_017029437.1 | XP_016884926.1 | 5 | 2461 | |||
XM_017029438.1 | XP_016884927.1 | 3 | 1137 | |||
GenBank mRNA | AK314134.1 | - | 2 | 978 | ||
BC069996.1 | - | 1 | 640 | 6971739 | ||
BC074722.2 | - | 1 | 604 | 30915172 | ||
BC095538.1 | - | 1 | 753 | 30706564 | ||
U90322.1 | - | 1 | 645 |