Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206739.1 | NP_001193668.1 | 7 | 607 | ||
NM_014288.4 | NP_055103.3 | 6 | 490 | |||
NR_045147.1 | 6 | 490 | ||||
XM_006710514.3 | XP_006710577.3 | 6 | 734 | |||
XM_006710515.3 | XP_006710578.3 | 6 | 734 | |||
XM_017000853.1 | XP_016856342.1 | 6 | 734 | |||
XM_017000854.1 | XP_016856343.1 | 6 | 525 | |||
XM_017000855.1 | XP_016856344.1 | 5 | 503 | |||
GenBank mRNA | AF175306.1 | - | 6 | 381 | ||
AK309622.1 | - | 6 | 533 | |||
AK312944.1 | - | 6 | 442 | |||
BC009929.2 | - | 6 | 397 | 3505579 | ||
BC014385.1 | - | 7 | 532 | 4064625 | ||
BU853146.1 | - | 6 | 495 | |||
U37139.1 | - | 6 | 480 |