Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SMUG1 SIRNA Assays ›
Gene Symbol:
SMUG1
Gene Name:
single-strand-selective monofunctional uracil-DNA glycosylase 1
Gene Aliases:
FDG, HMUDG, UNG3
Chromosome Location:
Chr.12: 54171815 - 54188994 on Build GRCh38
UniGene ID:
Hs.632721
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
21019
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SMUG1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001243787.1
NP_001230716.1
3
386
NM_001243788.1
NP_001230717.1
2
302
NM_001243789.1
NP_001230718.1
4
663
NM_001243790.1
NP_001230719.1
3
582
NM_001243791.1
NP_001230720.1
2
302
NM_014311.2
NP_055126.1
3
389
NR_045039.1
3
384
XM_006719319.3
XP_006719382.1
4
488
XM_006719320.3
XP_006719383.1
3
595
XM_006719321.3
XP_006719384.1
2
340
XM_006719322.3
XP_006719385.1
3
388
XM_011538109.2
XP_011536411.1
4
659
XM_011538110.2
XP_011536412.1
4
662
XM_011538111.2
XP_011536413.1
3
563
XM_011538112.2
XP_011536414.1
3
565
XM_011538113.2
XP_011536415.1
3
428
XM_011538114.2
XP_011536416.1
3
584
XM_011538115.2
XP_011536417.1
3
458
XM_011538116.2
XP_011536418.1
3
398
XM_011538117.2
XP_011536419.1
3
374
XM_011538118.2
XP_011536420.1
3
419
XM_011538119.2
XP_011536421.1
3
361
XM_011538120.2
XP_011536422.1
2
339
XM_011538121.2
XP_011536423.1
4
663
XM_011538122.2
XP_011536424.1
4
659
XM_011538123.2
XP_011536425.1
4
659
XM_017019113.1
XP_016874602.1
4
653
XM_017019114.1
XP_016874603.1
2
736
XM_017019115.1
XP_016874604.1
3
461
XM_017019116.1
XP_016874605.1
5
915
XM_017019117.1
XP_016874606.1
3
425
XM_017019118.1
XP_016874607.1
3
362
XM_017019119.1
XP_016874608.1
3
399
XM_017019120.1
XP_016874609.1
3
592
GenBank mRNA
AF125182.1
-
1
271
AK001235.1
-
3
368
AK001789.1
-
3
362
AK222473.1
-
3
365
AK290274.1
-
2
294
BC000417.2
-
4
609
2961516
BC088352.1
-
3
430
6470294
BC105607.1
-
2
282
6724466
BG706635.1
-
2
305
BG764502.1
-
4
473
BU195802.1
-
1
354
CD245413.1
-
1
266
DA430734.1
-
2
294
KU178578.1
-
1
239
KU178579.1
-
1
239
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
DNA glycosylase
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
base-excision repair
depyrimidination
oxidized pyrimidine nucleobase lesion DNA N-glycosylase activity
DNA binding
uracil DNA N-glycosylase activity
protein binding
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-6lbnp:80/10.239.161.162:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline