Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014177.2 | NP_054896.2 | 1 | 537 | ||
XM_005266689.3 | XP_005266746.1 | 1 | 521 | |||
GenBank mRNA | AF161503.1 | - | 1 | 438 | ||
AK289923.1 | - | 1 | 535 | |||
AK313936.1 | - | 1 | 525 | |||
BC000892.1 | - | 1 | 497 | 3463640 | ||
BG431414.1 | - | 1 | 564 |