Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DNAJC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014280.2 | NP_055095.2 | 2, 3 | 221 | ||
GenBank mRNA | AF083190.1 | - | 2, 3 | 228 | ||
AK300797.1 | - | 2, 3 | 210 | |||
BC033159.1 | - | 2, 3 | 206 | 4330676 | ||
BX538122.1 | - | 3, 4 | 338 | |||
CR456900.1 | - | 2, 3 | 221 | |||
DB294321.1 | - | 2, 3 | 221 |