Gene Symbol: ADAMTS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014243.2 | NP_055058.2 | 22 | 4330 | ||
XM_011532421.2 | XP_011530723.1 | 22 | 4362 | |||
XM_011532422.2 | XP_011530724.1 | 22 | 4861 | |||
GenBank mRNA | AB002364.1 | - | 22 | 4282 | ||
BC016451.1 | - | 5 | 1770 | |||
BC130287.1 | - | 22 | 4327 | 40146318 |