Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TMEM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278237.1 | NP_001265166.1 | 6 | 1103 | ||
NM_014254.2 | NP_055069.1 | 6 | 1093 | |||
XM_005268562.3 | XP_005268619.1 | 7 | 1203 | |||
XM_005268563.3 | XP_005268620.1 | 7 | 1276 | |||
XM_017018686.1 | XP_016874175.1 | 7 | 1158 | |||
XM_017018687.1 | XP_016874176.1 | 8 | 1320 | |||
GenBank mRNA | AB015633.1 | - | 6 | 1039 | ||
AK124211.1 | - | 2 | 1720 | |||
AK289382.1 | - | 6 | 987 | |||
BC002596.2 | - | 6 | 1024 | |||
BC013152.1 | - | 6 | 1025 | 3919181 | ||
BX647105.1 | - | 6 | 1092 | |||
BX647419.1 | - | 6 | 1092 |