Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014399.3 | NP_055214.1 | 4 | 577 | ||
GenBank mRNA | AF100759.1 | - | 4 | 497 | ||
AF120265.1 | - | 4 | 403 | |||
AK026587.1 | - | 4 | 541 | |||
AK075475.1 | - | 4 | 521 | |||
AK093487.1 | - | 4 | 538 | |||
AK316595.1 | - | 4 | 520 | |||
AY358995.1 | - | 4 | 508 | |||
BC033863.1 | - | 4 | 429 | 5187772 | ||
CR542107.1 | - | 4 | 335 |