Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DRD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000797.3 | NP_000788.2 | 1, 2 | 293 | ||
GenBank mRNA | EU432112.1 | - | 1, 2 | 293 | ||
L12398.1 | - | 1, 2 | 293 | |||
S76942.1 | - | 1, 2 | 280 |
Set Membership: |
G-protein coupled receptor |