Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000883.3 | NP_000874.2 | 7 | 603 | ||
NM_001102605.1 | NP_001096075.1 | 6 | 573 | |||
NM_001142573.1 | NP_001136045.1 | 4 | 373 | |||
NM_001142574.1 | NP_001136046.1 | 4 | 373 | |||
NM_001142576.1 | NP_001136048.1 | 6 | 504 | |||
NM_001304521.1 | NP_001291450.1 | 4 | 401 | |||
NM_183243.2 | NP_899066.1 | 5 | 495 | |||
XM_017012168.1 | XP_016867657.1 | 6 | 573 | |||
XM_017012169.1 | XP_016867658.1 | 5 | 495 | |||
XM_017012170.1 | XP_016867659.1 | 4 | 402 | |||
XM_017012171.1 | XP_016867660.1 | 6 | 691 | |||
XM_017012172.1 | XP_016867661.1 | 6 | 691 | |||
XM_017012173.1 | XP_016867662.1 | 6 | 573 | |||
GenBank mRNA | AK054667.1 | - | 5 | 495 | ||
AK092452.1 | - | 6 | 540 | |||
AK122994.1 | - | 6 | 572 | |||
AK131294.1 | - | 4 | 320 | |||
AK293413.1 | - | 4 | 345 | |||
AK297943.1 | - | 4 | 345 | |||
BC033622.1 | - | 5 | 486 | 5210847 | ||
BX406207.2 | - | 5 | 498 | |||
CD014008.1 | - | 5 | 287 | |||
CD014010.1 | - | 6 | 387 | |||
CR933672.1 | - | 539 | ||||
DA980797.1 | - | 5 | 495 | |||
DC299597.1 | - | 4 | 373 |