Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003215.2 | NP_003206.2 | 5 | 520 | ||
XM_011513737.2 | XP_011512039.1 | 5 | 501 | |||
XM_011513741.1 | XP_011512043.1 | 4 | 474 | |||
XM_017008569.1 | XP_016864058.1 | 4 | 1726 | |||
GenBank mRNA | BC101711.1 | - | 5 | 528 | 8069217 | |
BC101713.1 | - | 5 | 528 | 8069219 | ||
BC143487.1 | - | 5 | 528 | 9051995 | ||
D29767.1 | - | 5 | 546 |