Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206741.1 | NP_001193670.1 | 5 | 527 | ||
NM_005525.3 | NP_005516.1 | 4 | 525 | |||
NM_181755.2 | NP_861420.1 | 5 | 505 | |||
GenBank mRNA | AK313973.1 | - | 4 | 467 | ||
BC012593.1 | - | 4 | 474 | 4286028 | ||
BG619051.1 | - | 5 | 499 | |||
BX345837.2 | - | 4 | 438 | |||
CF596989.1 | - | 4 | 527 | |||
KJ534869.1 | - | 4 | 373 |