Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006581.3 | NP_006572.2 | 3 | 1163 | ||
XM_011535383.2 | XP_011533685.1 | 3 | 6900 | |||
XM_011535385.2 | XP_011533687.1 | 4 | 7017 | |||
XM_017010188.1 | XP_016865677.1 | 4 | 4164 | |||
XM_017010190.1 | XP_016865679.1 | 4 | 1090 | |||
GenBank mRNA | AB023021.1 | - | 3 | 1116 | ||
AJ238701.1 | - | 2 | 926 | |||
AK314966.1 | - | 3 | 1158 | |||
AY800264.1 | - | 1 | 822 | |||
BC036101.1 | - | 3 | 1140 | 5286309 |