Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TEN1-CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037709.1 | 10 | 2854 | |||
GenBank mRNA | AK097104.1 | - | 10 | 2811 | ||
BX647274.1 | - | 9 | 3422 |
Gene Symbol: CDK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258.2 | NP_001249.1 | 8 | 1094 | ||
GenBank mRNA | X66357.1 | - | 7 | 1109 |