Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SUV39H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282166.1 | NP_001269095.1 | 5 | 1387 | ||
NM_003173.3 | NP_003164.1 | 5 | 1140 | |||
GenBank mRNA | AF019968.1 | - | 5 | 1133 | ||
AK223071.1 | - | 5 | 1112 | |||
AK298233.1 | - | 6 | 846 | |||
AK299900.1 | - | 5 | 1387 | |||
AK312547.1 | - | 5 | 1112 | |||
BC006238.2 | - | 5 | 1131 | 3945632 | ||
CR541746.1 | - | 5 | 1088 | |||
L08238.1 | - | 4 | 1228 |