Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014438.4 | NP_055253.2 | 4 | 290 | ||
NM_173178.2 | NP_775270.1 | 4 | 290 | |||
XM_011510962.1 | XP_011509264.1 | 4 | 439 | |||
GenBank mRNA | AF200494.1 | - | 4 | 293 | ||
AF201833.1 | - | 4 | 293 | |||
BC101831.1 | - | 3 | 244 | 8069337 | ||
BC101833.1 | - | 3 | 244 | 8069339 |