Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: HERC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278186.1 | NP_001265115.1 | 18 | 2527 | ||
NM_015601.3 | NP_056416.2 | 20 | 2779 | |||
NM_022079.2 | NP_071362.1 | 21 | 2803 | |||
XM_011539592.2 | XP_011537894.1 | 22 | 2877 | |||
XM_011539593.2 | XP_011537895.1 | 21 | 2853 | |||
XM_011539594.2 | XP_011537896.1 | 23 | 2963 | |||
XM_011539595.2 | XP_011537897.1 | 23 | 3013 | |||
XM_011539596.2 | XP_011537898.1 | 22 | 2891 | |||
XM_011539597.2 | XP_011537899.1 | 20 | 2514 | |||
XM_017016041.1 | XP_016871530.1 | 19 | 2442 | |||
GenBank mRNA | AB046813.1 | - | 18 | 2657 | ||
AK027835.1 | - | 11 | 1346 | |||
AY221963.1 | - | 21 | 2740 | |||
AY650032.1 | - | 18 | 2464 | |||
BC015043.1 | - | 18 | 2430 | |||
BC039600.1 | - | 20 | 2689 | 5212048 | ||
BX537563.1 | - | 20 | 5266 |