Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HELZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014877.3 | NP_055692.2 | 13 | 1475 | ||
XM_005257888.4 | XP_005257945.1 | 13 | 1523 | |||
XM_005257889.4 | XP_005257946.1 | 13 | 1523 | |||
XM_006722214.3 | XP_006722277.1 | 13 | 1523 | |||
XM_006722215.3 | XP_006722278.1 | 11 | 1395 | |||
XM_006722216.3 | XP_006722279.1 | 2 | 225 | |||
XM_011525544.2 | XP_011523846.1 | 13 | 1594 | |||
XM_017025477.1 | XP_016880966.1 | 11 | 1395 | |||
XM_017025478.1 | XP_016880967.1 | 14 | 1657 | |||
GenBank mRNA | BC056895.1 | - | 12 | 1418 | ||
BC094881.1 | - | 11 | 1342 | 3076113 | ||
BC130582.1 | - | 13 | 1455 | 40146613 | ||
BC144083.1 | - | 13 | 1455 | 9052603 | ||
D29677.2 | - | 12 | 1433 |