Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002360.3 | NP_002351.1 | 3 | 986 | ||
XM_006715773.2 | XP_006715836.1 | 3 | 1039 | |||
GenBank mRNA | AF059194.1 | - | 3 | 986 | ||
AK092414.1 | - | 3 | 1793 | |||
BC143040.1 | - | 1 | 278 | |||
BC143041.1 | - | 1 | 280 | |||
BC148194.1 | - | 1 | 280 |