Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000913.5 | NP_000904.1 | 4 | 3110 | ||
NM_001200019.1 | NP_001186948.1 | 6 | 3218 | |||
NM_001318853.1 | NP_001305782.1 | 5 | 3261 | |||
NM_001318854.1 | NP_001305783.1 | 3 | 2824 | |||
NM_001318855.1 | NP_001305784.1 | 3 | 2867 | |||
NM_182647.3 | NP_872588.1 | 5 | 3261 | |||
XM_011528828.2 | XP_011527130.1 | 5 | 3763 | |||
XM_011528830.2 | XP_011527132.1 | 4 | 3498 | |||
XM_017027852.1 | XP_016883341.1 | 4 | 3280 | |||
XM_017027853.1 | XP_016883342.1 | 5 | 3200 | |||
XM_017027854.1 | XP_016883343.1 | 4 | 3135 | |||
XM_017027855.1 | XP_016883344.1 | 5 | 3227 | |||
GenBank mRNA | BC038433.1 | - | 6 | 3218 | 5164017 | |
CB048300.1 | - | 1 | 351 |
Set Membership: |
![]() |