Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016584.2 | NP_057668.1 | 3 | 456 | ||
XM_011538477.2 | XP_011536779.1 | 5 | 643 | |||
GenBank mRNA | AB030000.1 | - | 3 | 433 | ||
AF301620.1 | - | 3 | 440 | |||
AY359083.1 | - | 3 | 460 | |||
BC066267.1 | - | 3 | 323 | 6971848 | ||
BC066268.1 | - | 3 | 323 | 6971849 | ||
BC066269.1 | - | 3 | 323 | 6971851 | ||
BC067511.1 | - | 3 | 323 | 6971850 | ||
BC067512.1 | - | 3 | 323 | 6971852 | ||
BC067513.1 | - | 3 | 323 | 6971853 | ||
BM842783.1 | - | 3 | 456 |