Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL20RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278722.1 | NP_001265651.1 | 7 | 2405 | ||
NM_001278723.1 | NP_001265652.1 | 6 | 2545 | |||
NM_001278724.1 | NP_001265653.1 | 6 | 2479 | |||
NM_014432.3 | NP_055247.3 | 7 | 2724 | |||
XM_006715506.2 | XP_006715569.1 | 6 | 2121 | |||
XM_011535904.2 | XP_011534206.1 | 6 | 2232 | |||
XM_017010954.1 | XP_016866443.1 | 7 | 2913 | |||
XM_017010955.1 | XP_016866444.1 | 7 | 2373 | |||
GenBank mRNA | AF184971.1 | - | 7 | 2508 |