Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014439.3 | NP_055254.2 | 5 | 637 | ||
NM_173202.1 | NP_775294.1 | 4 | 574 | |||
NM_173203.1 | NP_775295.1 | 3 | 454 | |||
NM_173204.1 | NP_775296.1 | 4 | 517 | |||
NM_173205.1 | NP_775297.1 | 4 | 517 | |||
XM_011510963.2 | XP_011509265.1 | 6 | 2638 | |||
XM_011510964.2 | XP_011509266.1 | 5 | 2885 | |||
GenBank mRNA | AB451333.1 | - | 4 | 532 | ||
AB451478.1 | - | 4 | 532 | |||
AF167368.1 | - | 5 | 637 | |||
AF200496.1 | - | 5 | 643 | |||
AF201832.1 | - | 4 | 517 | |||
AF251118.1 | - | 5 | 667 | |||
AF251119.1 | - | 5 | 595 | |||
AF251120.1 | - | 4 | 475 | |||
AY071840.1 | - | 4 | 532 | |||
AY071841.1 | - | 3 | 412 | |||
BC020637.1 | - | 5 | 598 | 4731213 |