Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098.2 | NP_001089.1 | 10 | 1187 | ||
XM_017028812.1 | XP_016884301.1 | 10 | 1501 | |||
GenBank mRNA | AK293562.1 | - | 6 | 409 | ||
AK296260.1 | - | 11 | 1152 | |||
AK297213.1 | - | 10 | 1062 | |||
AK301321.1 | - | 5 | 328 | |||
AK302691.1 | - | 11 | 1137 | |||
AK314845.1 | - | 10 | 1195 | |||
BC014092.2 | - | 10 | 1187 | 4328775 | ||
BC026196.2 | - | 10 | 1190 | 5264179 | ||
CR456365.1 | - | 10 | 1179 | |||
CR536568.1 | - | 10 | 1165 | |||
GQ472196.1 | - | 10 | 1190 | |||
U80040.1 | - | 10 | 1185 |